N-Acetyl-L-leucine, 99%
N-Acetyl-L-leucine, 99%
N-Acetyl-L-leucine, 99%
Thermo Scientific Chemicals

N-Acetyl-L-leucine, 99%

CAS: 1188-21-2 | C8H15NO3 | 173.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
34701025025 gGlass bottle
Catalog number 347010250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Assay Percent Range99%
CAS Max %100.0
CAS Min %98.5
CAS1188-21-2
ChEBICHEBI:17786
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only