Flavin adenine dinucleotide disodium salt
Flavin adenine dinucleotide disodium salt
Flavin adenine dinucleotide disodium salt
Thermo Scientific Chemicals

Flavin adenine dinucleotide disodium salt

CAS: 84366-81-4 | C27H31N9Na2O15P2 | 829.52 g/mol
製品番号(カタログ番号)数量パッケージ
3472100101 gGlass bottle
製品番号(カタログ番号) 347210010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS84366-81-4
IUPAC Namedisodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[(2R,3S,4S)-5-{7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl}-2,3,4-trihydroxypentyl phosphonato]oxy}phosphonate
Molecular FormulaC27H31N9Na2O15P2
InChI KeyXLRHXNIVIZZOON-SJFBGTSINA-L
SMILES[Na+].[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1
さらに表示
UV spectrophotometry>=90 % (lambda max. 450 nm, pH 7)
Appearance (Form)Powder
UV spectrophotometrymolar absorptivity 11300 +/- 4%, on dry substance
Water=<10 % (K.F.)
Appearance (Color)Yellow to orange
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only