4-Bromo-2-fluorobenzoyl chloride, 99%
4-Bromo-2-fluorobenzoyl chloride, 99%
4-Bromo-2-fluorobenzoyl chloride, 99%
Thermo Scientific Chemicals

4-Bromo-2-fluorobenzoyl chloride, 99%

CAS: 151982-51-3 | C7H3BrClFO | 237.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 347460050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS151982-51-3
IUPAC Name4-bromo-2-fluorobenzoyl chloride
Molecular FormulaC7H3BrClFO
InChI KeyPCFIABOQFAFDAU-UHFFFAOYSA-N
SMILESFC1=CC(Br)=CC=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline mass or liquid
GC>=98.5 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only