3-Mercapto-1-hexanol, 96%
3-Mercapto-1-hexanol, 96%
3-Mercapto-1-hexanol, 96%
Thermo Scientific Chemicals

3-Mercapto-1-hexanol, 96%

CAS: 51755-83-0 | C6H14OS | 134.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 347560250
Price (USD)
258.00
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
258.00
Each
Chemical Identifiers
CAS51755-83-0
IUPAC Name3-sulfanylhexan-1-ol
Molecular FormulaC6H14OS
InChI KeyTYZFMFVWHZKYSE-UHFFFAOYNA-N
SMILESCCCC(S)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=95.0 %
Appearance (Color)Clear colorless
Refractive index1.4775 to 1.4835 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only