Product Image
Thermo Scientific™
96-Well Skirted PCR Plates
Ensure uniform sealing with these PCR plates, molded from 100% virgin polypropylene and featuring elevated rims.
Have Questions?
Catalog NumberColor
3482RRed
Catalog number 3482R
Price (MXN)
-
Color:
Red
Ensure uniform sealing with Thermo Scientific™ PCR plates, molded from 100% virgin polypropylene and featuring elevated rims.
  • Thermo Scientific™ PurePak™ packaging reduces contamination from reaching into a bulk bag of tubes by dividing tubes into ten separate PurePaks
  • Certified Rnase-, DNase- and pyrogen-free
Specifications
DescriptionSkirted
Product TypePCR Microplate
Skirt StyleSkirted
Array8 x 12
ColorRed
FormatStandard
MaterialPolypropylene
No. of Wells96
Unit SizeEach