Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Thermo Scientific Chemicals

Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™

Dichloromethane, 99.8%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, freon 30, methane dichloride, methane, dichloro, methylene bichloride, methylene chloride, methylene dichloride, narkotil, solaesthin, solmethine, 1L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless, >120 deg.C | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 348461000
Price (USD)
56.40
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
56.40
Each
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.8 %
Water=<0.005 % (Coulometric)
StabilizerApprox. 50 ppm Amylene
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only