Furfuryl glycidyl ether, 97%
Furfuryl glycidyl ether, 97%
Furfuryl glycidyl ether, 97%
Thermo Scientific Chemicals

Furfuryl glycidyl ether, 97%

CAS: 5380-87-0 | C8H10O3 | 154.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 348490250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5380-87-0
IUPAC Name2-{[(oxiran-2-yl)methoxy]methyl}furan
Molecular FormulaC8H10O3
InChI KeyRUGWIVARLJMKDM-UHFFFAOYNA-N
SMILESC(OCC1=CC=CO1)C1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4800 to 1.4820 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only