Castor oil, specified according to the requirements of Ph.Eur.
Castor oil, specified according to the requirements of Ph.Eur.
Castor oil, specified according to the requirements of Ph.Eur.
Thermo Scientific Chemicals

Castor oil, specified according to the requirements of Ph.Eur.

CAS: 8001-79-4 | C57H104O9 | 933.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 348850025
Price (USD)
307.65
Online exclusive
342.00
Save 34.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
307.65
Online exclusive
342.00
Save 34.35
Each
Chemical Identifiers
CAS8001-79-4
IUPAC Name1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
Molecular FormulaC57H104O9
InChI KeyZEMPKEQAKRGZGQ-UHFFFAOYNA-N
SMILESCCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Acid value=<1.5 mg KOH/g
Unsaponifiable matter=<0.8 %
Composition of fatty acids=<2.0 % (Palmitic acid)
Composition of fatty acids=<2.5 % (Stearic acid)
Composition of fatty acids2.5 to 6.0 % (Oleic acid)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only