Amber Screw Cap Micro Tubes
Amber Screw Cap Micro Tubes
Amber Screw Cap Micro Tubes
Amber Screw Cap Micro Tubes
Amber Screw Cap Micro Tubes
Thermo Scientific™

Amber Screw Cap Micro Tubes

Pure Protection: Prepare, transfer and store samples securely with Thermo Scientific Screw Cap Microtubes. Available in multiple volumes, packaging options and clean claims.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Bottom ShapeClosure TypeFit for Purpose
3488ANK2 mLSkirtedScrew Cap with O-ring (Unattached)PCR Ready
3422ANK0.5 mLSkirtedNot IncludedPCR Ready
3472ANK0.5 mLSkirtedScrew Cap with O-ring (Unattached)PCR Ready
3465ANK0.5 mLSkirtedAttached CapSterile and PCR Ready
3422ANKS0.5 mLSkirtedNot IncludedSterile and PCR Ready
3462ANK0.5 mLNon-skirtedNot IncludedPCR Ready
3460ANK0.5 mLNon-skirtedScrew Cap with O-ring (Unattached)PCR Ready
3431ANK0.5 mLNon-skirtedAttached CapSterile and PCR Ready
3462ANKS0.5 mLNon-skirtedNot IncludedSterile and PCR Ready
3422A0.5 mLSkirtedNot IncludedPCR Ready
3472A0.5 mLSkirtedScrew Cap with O-ring (Unattached)PCR Ready
3465A0.5 mLSkirtedAttached CapSterile and PCR Ready
3466ANK1.5 mLNon-skirtedNot IncludedPCR Ready
3464ANK1.5 mLNon-skirtedScrew Cap with O-ring (Unattached)PCR Ready
3461ANK1.5 mLNon-skirtedAttached CapSterile and PCR Ready
3466ANKS1.5 mLNon-skirtedNot IncludedSterile and PCR Ready
3478A1.5 mLSkirtedNot IncludedPCR Ready
3474A1.5 mLSkirtedScrew Cap with O-ring (Unattached)PCR Ready
3467A1.5 mLSkirtedAttached CapSterile and PCR Ready
3478AS1.5 mLSkirtedNot IncludedSterile and PCR Ready
3466A1.5 mLNon-skirtedNot IncludedPCR Ready
3464A1.5 mLNon-skirtedScrew Cap with O-ring (Unattached)PCR Ready
3466AS1.5 mLNon-skirtedNot IncludedSterile and PCR Ready
3490ANK2 mLSkirtedNot IncludedPCR Ready
3469ANK2 mLSkirtedAttached CapSterile and PCR Ready
3490ANKS2 mLSkirtedNot IncludedSterile and PCR Ready
3470ANK2 mLNon-skirtedNot IncludedPCR Ready
3468ANK2 mLNon-skirtedScrew Cap with O-ring (Unattached)PCR Ready
3463ANK2 mLNon-skirtedAttached CapSterile and PCR Ready
3470ANKS2 mLNon-skirtedNot IncludedSterile and PCR Ready
3490A2 mLSkirtedNot IncludedPCR Ready
3488A2 mLSkirtedScrew Cap with O-ring (Unattached)PCR Ready
3469A2 mLSkirtedAttached CapSterile and PCR Ready
3490AS2 mLSkirtedNot IncludedSterile and PCR Ready
3470A2 mLNon-skirtedNot IncludedPCR Ready
3468A2 mLNon-skirtedScrew Cap with O-ring (Unattached)PCR Ready
3463A2 mLNon-skirtedAttached CapSterile and PCR Ready
3470AS2 mLSkirtedNot IncludedSterile and PCR Ready
3460A0.5 mLConicalScrew Cap with O-ring (Unattached)PCR Ready
3431A0.5 mLConicalScrew Cap with O-ring (Attached)Sterile and PCR Ready
Catalog number 3488ANK
Price (MXN)
-
Capacity (Metric):
2 mL
Bottom Shape:
Skirted
Closure Type:
Screw Cap with O-ring (Unattached)
Fit for Purpose:
PCR Ready

Thermo Scientific™ Screw Cap Micro Tubes robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear externally threaded tubes and UV protective amber tubes. Options are available with or without a self-standing skirted bottom, non-knurl and knurled tubes for single hand operation, in 0.5mL, 1.5mL and 2.0mL sizes, and the option of multicolored caps, with or without tethered straps. Screw Cap Micro Tube Caps offer a durable design incorporating an O-ring that helps prevent leaks under demanding conditions.

  • Non-Sterile is free of Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor
  • Sterile E-Beam is free of Rnase/Dnase, Human and Mouse DNA, Pyrogen, Endotoxin, ATP, PCR Inhibitor
  • Packaging is tailored to meet broad requirements. Tubes are packaged with and without caps, caps are attached or unattached, placed in a single bag and double bagged feature for clean room environments is available upon request.
  • 100% virgin polypropylene resin specifically selected for high clarity and biologic applications
  • O-ring design, thickness and material selection have been optimized to provide protective features for maximum sealing ability, biologic interaction and temperature capability. The black colored O-ring is made from EPDM
  • Automated System Compatible. Whether automate is used for filling, capping or labeling the product is designed to withstand high torque pressures associated to automation. Product design helps to address common automated issues like: Fissure cracks, bowl feeder jam, over/under torqued caps and cross threading  
  • Volume range of 0.5, 1.5 and 2.0ml
  • Tube Color choices are an ultra-high clarity Natural color or the Amber option which provides ultra violet (UV) protection of 1.22% transmittance between 220 and 500nm
  • Standard or Tethered cap, all available in a rainbow of colors. Visit cap product page for additional information
  • Knurl tubes feature the ability to lock tube into specialty racks for single had operation
  • Non knurl tubes ideal for automation and labeling, tubes are plain removed of any labeling interference
  • Skirted or Self Standing tubes can stand upright on their own, no rack required
  • Conical tubes require a rack to support them in the upright position
  • Low nucleic acid and protein binding & low enzyme inhibition are features that Resin selection features
  • RCF range from 18,000 to 25,000 x g
  • Temperature Range -80°C to +95°C. Short term storage in -80°C, -20°C, dry ice and wet ice. Short term storage in vapor phase LN2. Suitable for a quick dip flash freeze in LN2
  • Water Bath compatible up to +95°C

Note: For screw caps, see Cat. No. 3471AS.

† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.

Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.

Not intended to submerge and store in liquid phase LN2.

Specifications
Binding PropertyLow Nucleic Acid and Protein Binding & Low Enzyme Inhibition
Clean ClaimsNon-Sterile Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor
IncludesNonKnurl
Max. TorqueManual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm
Packaging500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Product TypeScrew-cap Microtube
Temperature Range (Metric)Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage
UV Resistance(UV) Ultra violet protection avg. 1.22% transmittance between 220 and 500nm
AutoclavableNot Autoclavable
Bottom ShapeSkirted
Capacity (Metric)2 mL
Closure TypeScrew Cap with O-ring (Unattached)
ColorAmber
Fit for PurposePCR Ready
GraduatedYes
MaterialPolypropylene (Virgin)
Max. RCF25,000 x g
SterilityNon-sterile
Unit SizeCase of 5000