1,2-Dibromobenzene, 98%
1,2-Dibromobenzene, 98%
1,2-Dibromobenzene, 98%
Thermo Scientific Chemicals

1,2-Dibromobenzene, 98%

CAS: 583-53-9 | C6H4Br2 | 235.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 348931000
Price (TWD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS583-53-9
IUPAC Name1,2-dibromobenzene
Molecular FormulaC6H4Br2
InChI KeyWQONPSCCEXUXTQ-UHFFFAOYSA-N
SMILESBrC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.6100 to 1.6120 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow to light pink
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only