1-Naphthylacetic acid, 95%
1-Naphthylacetic acid, 95%
1-Naphthylacetic acid, 95%
Thermo Scientific Chemicals

1-Naphthylacetic acid, 95%

CAS: 86-87-3 | C12H10O2 | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 349640250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS86-87-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point126°C to 133°C
GC>=94 %
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only