tert-Butylperoxy isopropyl carbonate, 75% solution in aromatic free mineral spirit
tert-Butylperoxy isopropyl carbonate, 75% solution in aromatic free mineral spirit
tert-Butylperoxy isopropyl carbonate, 75% solution in aromatic free mineral spirit
Thermo Scientific Chemicals

tert-Butylperoxy isopropyl carbonate, 75% solution in aromatic free mineral spirit

CAS: 2372-21-6 | C8H16O4 | 176.212 g/mol
Have Questions?
Catalog NumberQuantity
349842500250 mL
Catalog number 349842500
Price (USD)
-
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS2372-21-6
CAS2372-21-6
SpecificationsSpecification SheetSpecification Sheet
Hydrolyzable chloride=<150 ppm (inorganic + organic)
Hydroperoxides=<1000 ppm (as TBHP)
Hydrolyzable chloride=<150 ppm (inorganic + organic)
Hydroperoxides=<1000 ppm (as TBHP)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only