Dicumyl peroxide, 99%
Dicumyl peroxide, 99%
Dicumyl peroxide, 99%
Thermo Scientific Chemicals

Dicumyl peroxide, 99%

CAS: 80-43-3 | C18H22O2 | 270.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 349885000
Price (USD)
186.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
186.00
Each
Chemical Identifiers
CAS80-43-3
IUPAC Name{2-[(2-phenylpropan-2-yl)peroxy]propan-2-yl}benzene
Molecular FormulaC18H22O2
InChI KeyXMNIXWIUMCBBBL-UHFFFAOYSA-N
SMILESCC(C)(OOC(C)(C)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point38°C to 41°C
Appearance (Form)Crystals
Infrared spectrumConforms
Titration Iodometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
RUO – Research Use Only