Cumyl hydroperoxide, 80%
Cumyl hydroperoxide, 80%
Cumyl hydroperoxide, 80%
Thermo Scientific Chemicals

Cumyl hydroperoxide, 80%

CAS: 80-15-9 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 349960010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS80-15-9
IUPAC Name2-phenylpropane-2-peroxol
Molecular FormulaC9H12O2
InChI KeyYQHLDYVWEZKEOX-UHFFFAOYSA-N
SMILESCC(C)(OO)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<450 APHA
Infrared spectrumConforms
Titration Iodometric80 to 85 %
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only