Pierce™ 10X Western Blot Transfer Buffer, Methanol-free
Thermo Scientific™

Pierce™ 10X Western Blot Transfer Buffer, Methanol-free

Thermo Scientific Pierce 10X Western Blot Transfer Buffer, Methanol-free is a space-saving stock solution for preparing the methanol-free transfer bufferRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
350405 L
35045500 mL
Catalog number 35040
Price (USD)
163.65
Online exclusive
182.00
Save 18.35
Each
Quantity:
5 L
Request bulk or custom format
Price (USD)
163.65
Online exclusive
182.00
Save 18.35
Each
Thermo Scientific Pierce 10X Western Blot Transfer Buffer, Methanol-free is a space-saving stock solution for preparing the methanol-free transfer buffer used with cooled, tank (wet) western blotting transfer apparatus.

Features of 10X Western Blot Transfer Buffer, Methanol-free:

• Transfer Buffer—diluted 10-fold in water, the solution is ready to use for electrophoresis (i.e., wet tank transfer from mini gels)
• Easy to use—no packets to open, no powder to dissolve, and no methanol required
• Increased accuracy—eliminates the possibility of powder remaining in a packet
• Saves time—10X concentration eliminates time spent waiting for powder to dissolve
• Saves space—storage as concentrated stock minimizes bench space needed for solutions

Pierce Concentrated Buffers are ready to use without having to weigh and dissolve dry ingredients or to adjust the pH with concentrated acid or base. Simply dilute the stock solution with pure water and proceed with your experiment. The 10X Western Blot Transfer Buffer is ready to use upon dilution to 1X with water; it does not contain methanol and the addition of methanol is not necessary for use in electrophoresis.

Applications:
• Wet (tank) western blot transfer of proteins to a membrane
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferTransfer Buffer
Concentration10X
Gel CompatibilitySDS-PAGE Gels
Quantity5 L
Shipping ConditionAmbient
Gel TypeSDS-PAGE
Product LinePierce
Product TypeWestern Blot Transfer Buffer
Unit SizeEach
Contents & Storage
Storage: Upon receipt store at 4°C, protected from moisture. Product is shipped at ambient temperature.

Frequently asked questions (FAQs)

Can you provide the shelf life for Pierce 10X Western Blot Transfer Buffer, Methanol-Free (Cat. No. 35045)?

Pierce 10X Western Blot Transfer Buffer is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.