Barium peroxide, 95%
Barium peroxide, 95%
Barium peroxide, 95%
Thermo Scientific Chemicals

Barium peroxide, 95%

CAS: 1304-29-6 | BaO2 | 169.33 g/mol
Have Questions?
產品號碼QuantityPackaging
35041025025 gPlastic bottle
產品號碼 350410250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS1304-29-6
規格Specification Sheet規格表
Appearance (Form)Powder
Appearance (Color)Beige
Titration Iodometric>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only