Thermo Scientific™
CU 65 Cooling Device
Related applications:
Anatomical Pathology
Stabilize temperature of the buffer solution in the buffer tray with the Thermo Scientific™ CU 65 Cooling Device, a special cooling unit only for the HM650V vibrating blade microtome.
Back to top
  • Tabletop unit
  • Temperature adjustable from ambient temperature down to -2°C for optimal preservation of tissue
  • Precise temperature setting via electronic temperature control
  • Integrated display shows temperature setting
  • Memory of selected temperature
  • Coated cooling element to be used in buffer tray
Product Specifications:
  • Electrical Requirements: 100-115V 50/60Hz/230-240V 50/60Hz
  • Dimensions (D x W x H): 54.6 x 102.4 x 50.8 in. (21.5 x 40.3 x 20cm)
  • Weight: 28.6 lb. (13kg)
Includes:
  • One bottle 50/50 ethylengycole/water solution
  • Instruction manual