GlutaMAX™ Supplement, 100 mL - FAQs
View additional product information for GlutaMAX™ Supplement - FAQs (35050038)
1 product FAQs found
What is the difference between Gibco GlutaMAX and glutamine supplements?
GlutaMAX Supplement is an improved cell culture supplement that can be used as a direct substitute for L-glutamine in your cell culture medium. Please go here to read more about the properties of GlutaMAX supplement (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/media-supplements/glutamax-media/glutamax-vs-glutamine.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.