4-chloro-5-nitro-o-phenylenediamine, 98%
4-chloro-5-nitro-o-phenylenediamine, 98%
4-chloro-5-nitro-o-phenylenediamine, 98%
Thermo Scientific Chemicals

4-chloro-5-nitro-o-phenylenediamine, 98%

CAS: 67073-39-6 | C6H6ClN3O2 | 187.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 350660050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS67073-39-6
IUPAC Name4-chloro-5-nitrobenzene-1,2-diamine
Molecular FormulaC6H6ClN3O2
InChI KeyLOQLMWFVXRZASN-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=C(C=C1N)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)Red
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only