Thermo Scientific™
Capitol Vial Temperature Indicator Strips
Related applications:
Industrial & Applied Science
Thermo Scientific™ Capitol Vial temperature indicator Strips are available separately for self application.
Back to top
  • Before use peel the strip off of the wax paper backing
  • Bright green or tan dot appears under temperature marking for you to accurately record the specimen temperature reading
  • Temperature measurement range 31.6° to 37.77°C (89° to 100°F)
  • Available in Celsius or Fahrenheit
Ordering Information:
Bag of 30