Methyl 3-fluorophenylacetate, 98%
Methyl 3-fluorophenylacetate, 98%
Methyl 3-fluorophenylacetate, 98%
Thermo Scientific Chemicals

Methyl 3-fluorophenylacetate, 98%

CAS: 64123-77-9 | C9H9FO2 | 168.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 351090050
Price (EUR)
23,50
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
23,50
Each
Chemical Identifiers
CAS64123-77-9
IUPAC Namemethyl 2-(3-fluorophenyl)acetate
Molecular FormulaC9H9FO2
InChI KeyOXYLCGRXCQARQV-UHFFFAOYSA-N
SMILESCOC(=O)CC1=CC=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.4870 to 1.4890 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only