ART™ Non-Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile
Thermo Scientific™
ART™ Non-Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile
The pipette tip and rack mail-back recycling program provides a simple and effective solution for recycling single use plastics. The TipCycle Recycle Kit arrives with your case of pipette tips to collect used pipette tips and empty racks. Attach the pre-paid shipping label and return for recycling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FormatTip StyleFit for Purpose
3511RC-05-HR10 μLRackLow Retention, Extended LengthSterile
3101RC-05-HR1000 μLHinged RackLow RetentionSterile
3101RC-HR1000 μLHinged RackExtended LengthSterile
3501RC-05-HR10 μLHinged RackLow RetentionSterile
3501RC-HR10 μLHinged RackStandardSterile
3521RC-05-HR10 μLRackLow Retention, Extended LengthCertified Quality
3521RC-HR20 μLHinged RackExtended LengthSterile
3551RC-05-HR200 μLHinged RackLow RetentionSterile
3551RC-HR200 μLHinged RackStandardSterile
3511RC-HR10 μLHinged RackExtended LengthSterile
Catalog number 3511RC-05-HR
Price (BRL)
-
Volume (Metric):
10 μL
Format:
Rack
Tip Style:
Low Retention, Extended Length
Fit for Purpose:
Sterile
Access tips with one hand using Thermo Scientific™ ART™ Non-filtered Hinged Rack Pipette Tips, designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover - now you have a traditional lift-off when you need it.
  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Provides: 1 case of pipette tips, instructions for return, a box liner and liner tie help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, FedEx return shipping label adhered to top of recycle package, then simply ship the recycle package
  • Complete selection of non-barrier and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • Sterile tips certified free: DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet. Secure snap design securely holds insert in place, preventing it from being lifted out while pipetting. Lid rim prevents racks from sliding around when stacked and carted around a lab.
Specifications
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Length (English)1.7 in.
Length (Metric)4.34 cm
ModelART 10 REACH
Packaging4800/Cs.
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Quantity4800/Cs.
AutoclavableNot Autoclavable
ColorNatural
FilteredNon-filtered
Fit for PurposeSterile
FormatRack
Product LineART
SterilitySterile
Tip StyleLow Retention, Extended Length
Volume (Metric)10 μL
Unit SizeCase of 4800