Toluene-d8, for NMR, 99.5 atom % D, with 0.03% TMS
Toluene-d8, for NMR, 99.5 atom % D, with 0.03% TMS
Toluene-d8, for NMR, 99.5 atom % D, with 0.03% TMS
Thermo Scientific Chemicals

Toluene-d8, for NMR, 99.5 atom % D, with 0.03% TMS

CAS: 2037-26-5 | C7H8 | 100.19 g/mol
Quantity:
5 mL
25 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 351500250
Price (JPY)
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS2037-26-5
IUPAC Name1-(²H₃)methyl(²H₅)benzene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-JGUCLWPXSA-N
SMILES[2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
D-Enrichment>=99.4 %
Water=<0.1 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only