ART™ Non-Filtered Standard Hinged Rack Pipette Tips
Thermo Scientific™
ART™ Non-Filtered Standard Hinged Rack Pipette Tips
Access tips one-handed with this versatile hinged rack, which is designed for the way you work.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip StyleFor Use With (Equipment)Fit for PurposeUnit Size
3521-05-HR20 μLLow RetentionEppendorfSterileCase of 4800
3101-05-HR1000 μLLow RetentionUniversal FitSterileCase of 3072
3102-05-HR1000 μLLow RetentionUniversal FitCertified QualityCase of 3072
3101-HR1000 μLStandardUniversal FitSterileCase of 3072
3102-HR1000 μLStandardUniversal FitCertified QualityCase of 3072
3771-05-HR300 μLLow RetentionUniversal FitSterileCase of 4800
3772-05-HR300 μLLow RetentionUniversal FitCertified QualityCase of 4800
3772-05-HRPK
also known as 3772-05-HR
300 μLLow RetentionUniversal FitCertified QualityPack of 960
3772-HR300 μLStandardUniversal FitCertified QualityCase of 4800
3771-HR300 μLStandardUniversal FitSterileCase of 4800
3771-HRPK300 μLStandardUniversal FitSterilePack of 960
3551-05-HR200 μLLow RetentionUniversal FitSterileCase of 4800
3552-05-HR200 μLLow RetentionUniversal FitCertified QualityCase of 4800
3551-HR200 μLStandardUniversal FitSterileCase of 4800
3552-HR200 μLStandardUniversal FitCertified QualityCase of 4800
3522-05-HR20 μLLow RetentionEppendorfCertified QualityCase of 4800
3522-05-HRPK
also known as 3522-05-HR
20 μLLow RetentionEppendorfCertified QualityPack of 960
3522-HR20 μLStandardEppendorfCertified QualityCase of 4800
3511-HR10 μLExtended LengthUniversal FitSterileCase of 4800
3501-05-HR10 μLLow RetentionUniversal FitSterileCase of 4800
3502-05-HR10 μLLow RetentionUniversal FitCertified QualityCase of 4800
3501-HR10 μLStandardUniversal FitSterileCase of 4800
3502-HR10 μLStandardUniversal FitCertified QualityCase of 4800
3521-HR20 μLStandardEppendorfSterileCase of 4800
Catalog number 3521-05-HR
Price (CLP)
-
Volume (Metric):
20 μL
Tip Style:
Low Retention
For Use With (Equipment):
Eppendorf
Fit for Purpose:
Sterile
Unit Size:
Case of 4800
Access tips with one hand using Thermo ScientificTM ARTTM Nonfiltered Hinged Rack Pipette Tips, which is designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover--now you have a traditional lift-off when you need it.
Features:
  • Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • Sterile tips are DNase, RNase, DNA, ATP, and Endotoxin Free
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
Specifications
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
DescriptionART Non-filtered, LR, Sterile, Hinged Rack
Length (English)1.8 in.
Length (Metric)4.57 cm
ModelART Non-filtered
ColorNatural
FilteredNon-filtered
Fit for PurposeSterile
For Use With (Equipment)Eppendorf
FormatHinged Rack
Product LineART
SterilitySterile
Tip StyleLow Retention
Volume (Metric)20 μL
Unit SizeCase of 4800