ART™ Non-Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile
Thermo Scientific™

ART™ Non-Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile

The pipette tip and rack mail-back recycling program provides a simple and effective solution for recycling single use plastics. The TipCycle Recycle Kit arrives with your case of pipette tips to collect used pipette tips and empty racks. Attach the pre-paid shipping label and return for recycling.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoVolumen (métrico)FormatoEstilo de puntaFit for Purpose
3521RC-05-HR20 μLGradillaRetención baja, Extended LengthCertificado de calidad
3101RC-05-HR10 μLGradilla con bisagrasRetención bajaEstéril
3101RC-HR1000 μLGradilla con bisagrasEstándarEstéril
3501RC-05-HR1000 μLGradilla con bisagrasRetención bajaEstéril
3501RC-HR1000 μLGradilla con bisagrasEstándarEstéril
3511RC-05-HR10 μLGradillaRetención baja, Extended LengthEstéril
3521RC-HR20 μLGradilla con bisagrasEstándarEstéril
3551RC-05-HR200 μLGradilla con bisagrasRetención bajaEstéril
3551RC-HR200 μLGradilla con bisagrasEstándarEstéril
3511RC-HR10 μLGradilla con bisagrasEstándarEstéril
Número de catálogo 3521RC-05-HR
Precio (CLP)
-
Volumen (métrico):
20 μL
Formato:
Gradilla
Estilo de punta:
Retención baja, Extended Length
Fit for Purpose:
Certificado de calidad

Access tips with one hand using Thermo Scientific™ ART™ Non-filtered Hinged Rack Pipette Tips, designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover - now you have a traditional lift-off when you need it.

  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Provides: 1 case of pipette tips, instructions for return, a box liner and liner tie help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, FedEx return shipping label adhered to top of recycle package, then simply ship the recycle package
  • Complete selection of non-barrier and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • Sterile tips certified free: DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet. Secure snap design securely holds insert in place, preventing it from being lifted out while pipetting. Lid rim prevents racks from sliding around when stacked and carted around a lab.
Especificaciones
Declaraciones de limpiezaSin ADNasa, ARNasa, ADN, ATP, inhibidor de PCR ni endotoxinas
Incluye1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Longitud (imperial)1,7 in
Longitud (métrico)4.34 cm
ModeloART 20 LR
Envase4800/Cs.
Tipo de productoTipCycle, Pipette Tip and Rack Recycling Kit
Esterilizables en autoclaveNo Autoclavable
ColorNatural
filtradas porSin filtro
Fit for PurposeCertificado de calidad
FormatoGradilla
Línea de productosART
EsterilidadEstéril
Estilo de puntaRetención baja, Extended Length
Volumen (métrico)20 μL
Unit SizeCase of 4800