ART™ Reload System
Thermo Scientific™
ART™ Reload System
Reduce waste and save benchtop and storage space with this advanced nonfiltered reload system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)For Use With (Equipment)ModelFit for PurposeUnit Size
3522-05-RTPK
also known as 3522-05-RT
20 μLEppendorfART Non-filteredCertified QualityPack of 960
3101-05-RT1000 μLUniversal FitART Non-filteredSterileCase of 2800
3101-05-RTPK
also known as 3101-05-RT
1000 μLUniversal FitART Non-filteredSterilePack of 560
3522-RTPK
also known as 3522-RT
20 μLEppendorfART Ultra Micro Non-FilteredCertified QualityPack of 960
3101-RT1000 μLUniversal FitART Non-filteredSterileCase of 2800
3102-05-RT1000 μLUniversal FitART Non-filteredCertified QualityCase of 2800
3102-05-RTPK
also known as 3102-05-RT
1000 μLUniversal FitART Non-filteredCertified QualityPack of 560
3501-05-RT10 μLUniversal FitART Non-filteredSterileCase of 4800
3501-RT10 μLUniversal FitART Non-filteredSterileCase of 4800
3501-RTPK10 μLUniversal FitART Non-filteredSterilePack of 960
3502-05-RT10 μLUniversal FitART Non-filteredCertified QualityCase of 4800
3502-05-RTPK
also known as 3502-05-RT
10 μLUniversal FitART Non-filteredCertified QualityPack of 960
3502-RT10 μLUniversal FitART Ultra Micro Non-FilteredCertified QualityCase of 4800
3521-05-RT20 μLEppendorfART Non-filteredSterileCase of 4800
3521-RT20 μLEppendorfART Non-filteredSterileCase of 4800
3521-RTPK
also known as 3521-RT
20 μLEppendorfART Non-filteredSterilePack of 960
3522-05-RT20 μLEppendorfART Non-filteredCertified QualityCase of 4800
3522-RT20 μLEppendorfART Non-filteredCertified QualityCase of 4800
3551-05-RT200 μLUniversal FitART Non-filteredSterileCase of 4800
3551-RT200 μLUniversal FitART Non-filteredSterileCase of 4800
3552-05-RT200 μLUniversal FitART Non-filteredCertified QualityCase of 4800
3552-RT200 μLUniversal FitART Non-filteredCertified QualityCase of 4800
3771-05-RT300 μLUniversal FitART Non-filteredSterileCase of 4800
3771-05-RTPK
also known as 3771-05-RT
300 μLUniversal FitART Non-filteredSterilePack of 960
3771-RT300 μLUniversal FitART Non-filteredSterileCase of 4800
3771-RTPK
also known as 3771-RT
300 μLUniversal FitART Non-filteredSterilePack of 960
3772-05-RT300 μLUniversal FitART Non-filteredCertified QualityCase of 4800
3772-05-RTPK
also known as 3772-05-RT
300 μLUniversal FitART Non-filteredCertified QualityPack of 960
3772-RT300 μLUniversal FitART Non-filteredCertified QualityCase of 4800
3772-RTPK
also known as 3772-RT
300 μLUniversal FitART Non-filteredCertified QualityPack of 960
Catalog number 3522-05-RTPK
also known as 3522-05-RT
Price (MXN)
-
Volume (Metric):
20 μL
For Use With (Equipment):
Eppendorf
Model:
ART Non-filtered
Fit for Purpose:
Certified Quality
Unit Size:
Pack of 960
Reduce waste and save benchtop and storage space with the advanced Thermo Scientific™ ART™ Nonfiltered Reload System. This system provides an environmentally responsible option without compromising sterility.
Features:
  • Stackable barrier tip packaging allows for tips to pass through offset holes in lower insert
  • Blister sleeve guide rails align tip inserts for smooth transfer into the rack
  • DNase, RNase, DNA, ATP, and Endotoxin Free
  • Outer sleeve protects tips from exterior contamination while refilling inserts
  • Tip packaging option reduces plastic waste
  • Rigid insert design provides reliable tip loading
  • Lot number and description are printed on every insert
Note:
Empty rack required for -RT parts
Specifications
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
Length (English)1.8 in.
Length (Metric)4.57 cm
ModelART Non-filtered
Quantity4800/Cs.
ColorNatural
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Eppendorf
FormatRack
Product LineART
SterilityNon-sterile
Tip StyleLow Retention
Volume (Metric)20 μL
Unit SizePack of 960