Search
Search
| Catalog Number | Holds | Product Type | Dimensions (L x W) | For Use With (Equipment) |
|---|---|---|---|---|
| 3523-8 | 4 x 1 L Erlenmeyer Flask | Erlenmeyer Flask | 11 x 13 in. | MaxQ 2000, 2506, 2508, 4450 |
| 3523-1 | 60 x 10 mL Erlenmeyer Flask | Erlenmeyer Flask | 11 x 13 in. | MaxQ 2000, 2506, 2508, 4450 |
| 3523-4 | 15 x 1125 mL Erlenmeyer Flasks | Erlenmeyer Flask | 11 x 13 in. | MaxQ 2000, 2506, 2508, 4450 |
| 3523-46 | Tray | Shaker Platform Utility Tray | 11 x 13 in. | MaxQ 2000, 2506, 2508, 4450 |
| 3523-49 | Adjustable Vessel Platform | 11 x 13 in. | MaxQ 2000, 2506, 2508 | |
| 3523-5 | 10 x 250 Erlenmeyer Flasks | Erlenmeyer Flask | 11 x 13 in. | MaxQ 2000, 2506, 2508, 4450 |
| 3523-51 | Platelet bag platform | 11 x 13 in. | MaxQ 2000, 2506, 2508 | |
| 3523-52 | Adjustable Vessel Platform | 18 x 18 in. | MaxQ 2000, 2506, 2508 | |
| 3523-7 | 6 x 500 mL Erlenmeyer Flasks | Erlenmeyer Flask | 11 x 13 in. | MaxQ 2000, 2506, 2508, 4450 |
| 3523-9 | 4 x 2 L Erlenmeyer Flasks | Erlenmeyer Flask | 11 x 13 in. | MaxQ 2000, 2506, 2508 |
| 3524-2 | 64 x 25 mL Erlenmeyer Flasks | Erlenmeyer Flask | 18 x 18 in. | MaxQ 2000, 2506, 2508, 4000, 6000 |
| 3524-3 | 64 x 50 mL Erlenmeyer Flasks | Erlenmeyer Flask | 18 x 18 in. | MaxQ 2000, 2506, 2508, 4000, 6000 |
| 3524-38 | 44 x 100 mL Beaker | Beaker | 18 x 18 in. | MaxQ 2000, 2506, 2508, 4000, 6000 |
| 3524-4 | Erlenmeyer Flask | 18 x 18 in. | MaxQ 2000, 2506, 2508, 4000, 6000 | |
| 3524-41 | 16 x 1600 mL Beakers | Beaker | 18 x 18 in. | MaxQ 2000, 2506, 2508, 4000, 6000 |
| 3524-46 | Utility Tray Platform | 18 x 18 in. | MaxQ 2000, 2506, 2508, 4000, 6000 | |
| 3524-49 | 500 mL to 2 L Separatory Funnel | Separatory Funnel | 18 x 18 in. | MaxQ 2000, 2506, 2508, 4000, 6000 |
| 3524-6 | 16 x 1300 mL Erlenmeyer Flask | Erlenmeyer Flask | 18 x 18 in. | MaxQ 2000, 2506, 2508, 4000, 6000 |
| 3524-60 | Adjustable Vessel Platform | 18 x 18 in. | MaxQ 4000, 6000 | |
| 3524-7 | 16 x 1500 mL Erlenmeyer Flask | Erlenmeyer Flask | 18 x 18 in. | MaxQ 2000, 2506, 2508, 4000, 6000 |
| 3524-8 | 9 x 1 L Erlenmeyer Flask | Erlenmeyer Flask | 18 x 18 in. | MaxQ 2000, 2506, 2508, 4000, 6000 |
| 3524-9 | 5 x 2 L Erlenmeyer Flask | Erlenmeyer Flask | 18 x 18 in. | MaxQ 2000, 2506, 2508, 4000, 6000 |
Pick the optimal choice for a busy lab with Thermo Scientific™ Dedicated Platforms for MaxQ™ 2000/2506/2508/4000/4450/6000 Shakers. MaxQ platforms are model-specific and come with pre-installed clamps to maximize capacity. MaxQ shakers, platforms and accessories provide exceptional innovation, reliability and accuracy.