Methyl sulfoxide-d6, for NMR, with 0.03% TMS, in 0.75 ml ampoules, 99.9 atom% D
Methyl sulfoxide-d6, for NMR, with 0.03% TMS, in 0.75 ml ampoules, 99.9 atom% D
Methyl sulfoxide-d6, for NMR, with 0.03% TMS, in 0.75 ml ampoules, 99.9 atom% D
Thermo Scientific Chemicals

Methyl sulfoxide-d6, for NMR, with 0.03% TMS, in 0.75 ml ampoules, 99.9 atom% D

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Catalog NumberQuantity
35254007510 x 0.75 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 352540075
Price (USD)
-
Quantity:
10 x 0.75 mL
Request bulk or custom format
Chemical Identifiers
CAS2206-27-1
IUPAC Name(²H₃)methanesulfinyl(²H₃)methane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
D-Enrichment>=99.85 %
Water=<0.03 % (Coulometric)
TMS0.02 to 0.04 v/v %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only