Pyridoxal hydrochloride, 99%
Pyridoxal hydrochloride, 99%
Pyridoxal hydrochloride, 99%
Thermo Scientific Chemicals

Pyridoxal hydrochloride, 99%

CAS: 65-22-5 | C8H9NO3·HCl | 203.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 352710010
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.60
Save 4.95
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.60
Save 4.95
Each
Chemical Identifiers
CAS65-22-5
IUPAC Namehydrogen 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde chloride
Molecular FormulaC8H10ClNO3
InChI KeyFCHXJFJNDJXENQ-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC1=NC=C(CO)C(C=O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point171°C to 175°C (Dec)
Titration with HClO4>=98.5 %
Appearance (Color)White to yellow to light beige
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only