Orotic acid, 98%, anhydrous
Orotic acid, 98%, anhydrous
Orotic acid, 98%, anhydrous
Thermo Scientific Chemicals

Orotic acid, 98%, anhydrous

CAS: 65-86-1 | C5H4N2O4 | 156.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 352880100
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
45.20
Save 6.55
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
45.20
Save 6.55
Each
Chemical Identifiers
CAS65-86-1
IUPAC Name2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
Molecular FormulaC5H4N2O4
InChI KeyPXQPEWDEAKTCGB-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=O)NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Water=<2 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only