Dibutyl maleate, 97%
Dibutyl maleate, 97%
Dibutyl maleate, 97%
Thermo Scientific Chemicals

Dibutyl maleate, 97%

CAS: 105-76-0 | C12H20O4 | 228.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 353102500
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.10
Save 4.45
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.10
Save 4.45
Each
Chemical Identifiers
CAS105-76-0
IUPAC Name1,4-dibutyl (2Z)-but-2-enedioate
Molecular FormulaC12H20O4
InChI KeyJBSLOWBPDRZSMB-FPLPWBNLSA-N
SMILESCCCCOC(=O)\C=C/C(=O)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4440 to 1.4460 (20°C, 589 nm)
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only