1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
Thermo Scientific Chemicals

1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%

CAS: 174501-64-5 | C8H15F6N2P | 284.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 354170050
Price (USD)
49.50
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
49.50
Each
Chemical Identifiers
CAS174501-64-5
IUPAC Name1-butyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC8H15F6N2P
InChI KeyIXQYBUDWDLYNMA-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
HPLC>=98.0 %
Water=<1.0 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquid
RUO – Research Use Only