ART™ Non-Filtered Extended Length Hinged Rack Pipette Tips
Actual product may vary
ART™ Non-Filtered Extended Length Hinged Rack Pipette Tips
Access tips one-handed with this versatile hinged rack, designed for the way you work.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityTip StyleFit for Purpose
3542-HR200 μLNon-sterileUniversal FitCertified Quality
3512-HR10 μLNon-sterileUniversal FitCertified Quality
3512-05-HR10 μLNon-sterileLow RetentionCertified Quality
3511-05-HR10 μLSterileLow RetentionSterile
3541-HR200 μLSterileUniversal FitSterile
3542-05-HR200 μLNon-sterileLow RetentionCertified Quality
3541-05-HR200 μLSterileLow RetentionSterile
3792-HR1000 μLNon-sterileUniversal FitCertified Quality
3792-05-HR1000 μLNon-sterileLow RetentionCertified Quality
3791-HR1000 μLSterileUniversal FitSterile
3152-HR1250 μLNon-sterileUniversal FitCertified Quality
3151-HR1250 μLSterileUniversal FitSterile
3152-05-HR1250 μLNon-sterileLow RetentionCertified Quality
3151-05-HR1250 μLSterileLow RetentionSterile
Catalog number 3542-HR
HSN 84799090
Price (INR)
10,485.00
PK
Volume (Metric):
200 μL
Sterility:
Non-sterile
Tip Style:
Universal Fit
Fit for Purpose:
Certified Quality
Price (INR)
10,485.00
PK
Access tips with one hand using Thermo Scientific™ ART™ Non-Filtered Extended Length Hinged Rack Pipette Tips, designed for the way you work. No longer worry about your tips spilling on your bench or the floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover — now you have a traditional lift-off when you need it.
Features:
  • Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • DNase, RNase, DNA, ATP, and Endotoxin Free
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
Specifications
DescriptionART Non-Filtered, Non-sterile, Rack
Volume (Metric)200 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
ModelART Non-filtered
Length (English)3.6 in.
Length (Metric)9.09cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatRack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeCertified Quality
Unit SizePK