Methanephosphonic acid, 98%, Thermo Scientific Chemicals
Methanephosphonic acid, 98%, Thermo Scientific Chemicals
Methanephosphonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanephosphonic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 354330250
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS993-13-5
IUPAC Namemethylphosphonic acid
Molecular FormulaCH5O3P
InChI KeyYACKEPLHDIMKIO-UHFFFAOYSA-N
SMILESCP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Melting point103°C to 107°C
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only