1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol, 99.8%, for peptide synthesis

CAS: 920-66-1 | C3H2F6O | 168.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
354861000100 mLGlass bottle
Catalog number 354861000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS920-66-1
IUPAC Name1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular FormulaC3H2F6O
InChI KeyBYEAHWXPCBROCE-UHFFFAOYSA-N
SMILESOC(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.02 % (K.F.)
Free amine=<0.0002 %
Appearance (Form)Clear liquid
GC>=99.8 %
Residue after evaporation=<0.0005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only