1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis

CAS: 872-50-4 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 354900010
Price (USD)
170.65
Online exclusive
189.00
Save 18.35
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
170.65
Online exclusive
189.00
Save 18.35
Each
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Free amine=<0.01 %
Water=<0.1 % (K.F.)
Appearance (Form)Clear liquid
GC>=99.4 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only