Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Thermo Scientific™

Cryo Vial Closure Color Coders

Green features
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct Line
355018GreenNunc
354755WhiteNunc
354879BlueNunc
354968RedNunc
355077YellowNunc
355158OrangeNunc
375868BrownNunc
375884PinkNunc
375906GrayNunc
375922PurpleNunc
375930AssortedNunc
5045-0003BlueNalgene
5045-0000WhiteNalgene
5045-0002YellowNalgene
5045-0004GreenNalgene
5045-0005RedNalgene
Catalog number 355018
Price (MXN)
-
Color:
Green
Product Line:
Nunc
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.
  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.
Specifications
DescriptionNunc™ CryoColor Vial Closure Coder, PS
For Use With (Equipment)Cryogenic Vial
Product TypeVial Closure
ColorGreen
MaterialPolystyrene
Product LineNunc
Quantity500/Pk., 2000/Cs.
Unit SizeCase of 2000

Frequently asked questions (FAQs)

Are your Nunc CryoColor Vial Closure Coders removable?

Once inserted, the Nunc CryoColor Vial Closure Coders cannot be removed.