2,7-Dibromofluorene, 99%
2,7-Dibromofluorene, 99%
2,7-Dibromofluorene, 99%
Thermo Scientific Chemicals

2,7-Dibromofluorene, 99%

CAS: 16433-88-8 | C13H8Br2 | 324.02 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 355240050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16433-88-8
IUPAC Name2,7-dibromo-9H-fluorene
Molecular FormulaC13H8Br2
InChI KeyAVXFJPFSWLMKSG-UHFFFAOYSA-N
SMILESBrC1=CC=C2C(CC3=CC(Br)=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow
Melting point164°C to 167°C
Appearance (Form)Crystalline powder or crystals
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only