Potassium superoxide
Potassium superoxide
Potassium superoxide
Thermo Scientific Chemicals

Potassium superoxide

CAS: 12030-88-5 | KO2 | 71.1 g/mol
数量:
25 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 355420250
価格(JPY)
-
数量:
25 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12030-88-5
IUPAC Namepotassium dioxygen
Molecular FormulaKO2
InChI KeyCLSKHAYBTFRDOV-UHFFFAOYSA-N
SMILES[K+].O=O
さらに表示
Oxygen (O)>=215 cm³/g O2-content
Appearance (Color)Yellow
Appearance (Form)Powder or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only