Phorbol 12-myristate 13-acetate, 97%
Phorbol 12-myristate 13-acetate, 97%
Phorbol 12-myristate 13-acetate, 97%
Thermo Scientific Chemicals

Phorbol 12-myristate 13-acetate, 97%

CAS: 16561-29-8 | C36H56O8 | 616.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Packaging:
Glass bottle
Catalog number 356150010
Price (EUR)
90,65
Online exclusive
101,00
Save 10,35
Each
Quantity:
1 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
90,65
Online exclusive
101,00
Save 10,35
Each
Chemical Identifiers
CAS16561-29-8
IUPAC Name(1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular FormulaC36H56O8
InChI KeyPHEDXBVPIONUQT-RGYGYFBISA-N
SMILESCCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Color)Colorless to white to light yellow
Appearance (Form)Film or solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only