Dimethyl 5-hydroxyisophthalate, 99%
Dimethyl 5-hydroxyisophthalate, 99%
Dimethyl 5-hydroxyisophthalate, 99%
Thermo Scientific Chemicals

Dimethyl 5-hydroxyisophthalate, 99%

CAS: 13036-02-7 | C10H10O5 | 210.19 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
產品號碼 356440250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS13036-02-7
IUPAC Name1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate
Molecular FormulaC10H10O5
InChI KeyDOSDTCPDBPRFHQ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(=CC(O)=C1)C(=O)OC
檢視更多
規格Specification Sheet規格表
Melting point162°C to 165°C
Appearance (Color)White to light beige
Appearance (Form)Powder and/or lumps
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only