Bis(neopentyl glycolato)diboron, 98%
Bis(neopentyl glycolato)diboron, 98%
Bis(neopentyl glycolato)diboron, 98%
Thermo Scientific Chemicals

Bis(neopentyl glycolato)diboron, 98%

CAS: 201733-56-4 | C10H20B2O4 | 225.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 358890010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS201733-56-4
IUPAC Name2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
Molecular FormulaC10H20B2O4
InChI KeyMDNDJMCSXOXBFZ-UHFFFAOYSA-N
SMILESCC1(C)COB(OC1)B1OCC(C)(C)CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
GC>=97.5 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only