3-Chlorophenylboronic acid, 97%
3-Chlorophenylboronic acid, 97%
3-Chlorophenylboronic acid, 97%
Thermo Scientific Chemicals

3-Chlorophenylboronic acid, 97%

CAS: 63503-60-6 | C6H6BClO2 | 156.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 359000010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS63503-60-6
IUPAC Name(3-chlorophenyl)boronic acid
Molecular FormulaC6H6BClO2
InChI KeySDEAGACSNFSZCU-UHFFFAOYSA-N
SMILESOB(O)C1=CC(Cl)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow-green
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only