2,4-Difluorophenylboronic acid, 98%
2,4-Difluorophenylboronic acid, 98%
2,4-Difluorophenylboronic acid, 98%
Thermo Scientific Chemicals

2,4-Difluorophenylboronic acid, 98%

CAS: 144025-03-6 | C6H5BF2O2 | 157.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 359130100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS144025-03-6
IUPAC Name(2,4-difluorophenyl)boronic acid
Molecular FormulaC6H5BF2O2
InChI KeyQQLRSCZSKQTFGY-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point247°C to 251°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only