ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips
Thermo Scientific™

ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips

Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFormat
35921000 μLNon-sterileLift-off Lid Rack
3531200 μLSterileLift-off Lid Rack
35901000 μLNon-sterileBulk
35911000 μLSterileRack
Catalog number 3592
Price (MXN)
-
Volume (Metric):
1000 μL
Sterility:
Non-sterile
Format:
Lift-off Lid Rack
Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows. The wide orifice prevents shearing of cells and DNA and RNA creating longer, continuous strands. The wide opening also allows for easier pipetting of viscous fluids or solutions. Ideal for use in reference labs and blood banking applications.
  • Wide orifice is larger than standard tip openings making it ideal for pipetting fragile cell lines and viscous samples
  • Ideal for genomic work
  • Universal Tip, fits most brands of pipettes
  • ART™ non-filtered tips deliver the same proven quality, performance and reliability as ART™ barrier
  • Non-filtered tip
  • ART™ tips are DNase, RNase, DNA, ATP, PCR, and Endotoxin Free
  • Lift-off lid racks provide access to all tips, great for multichannel pipetting
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, Bioburden, PCR Inhibitor, Endotoxin, and Pyrogen Free
DescriptionART 1000G, Non-filtered, non-sterile, Racked, Lift-off Lid
DisposableSingle-use
FeaturesWide bore orifice
For Use With (Application)Genomic workflows, fragile cell lines, viscous liquids
ModelART 1000G
Packaging3200/Pk.
Quantity3200/Pk.
AutoclavableAutoclavable
ColorClear
FilteredNon-filtered
For Use With (Equipment)Universal Fit
FormatLift-off Lid Rack
MaterialVirgin Polypropylene
Product LineART
SterilityNon-sterile
Tip StyleWide Orifice
Volume (Metric)1000 μL
Unit SizeCase of 3200