4-Formylphenylboronic acid, 97%
4-Formylphenylboronic acid, 97%
4-Formylphenylboronic acid, 97%
Thermo Scientific Chemicals

4-Formylphenylboronic acid, 97%

CAS: 87199-17-5 | C7H7BO3 | 149.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 359380010
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
35.70
Save 5.05
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
35.70
Save 5.05
Each
Chemical Identifiers
CAS87199-17-5
IUPAC Name(4-formylphenyl)boronic acid
Molecular FormulaC7H7BO3
InChI KeyVXWBQOJISHAKKM-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Appearance (Form)Powder or crystals or flakes and/or chunks
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only