5-Indolylboronic acid, 97%
5-Indolylboronic acid, 97%
5-Indolylboronic acid, 97%
Thermo Scientific Chemicals

5-Indolylboronic acid, 97%

CAS: 144104-59-6 | C8H8BNO2 | 160.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 359465000
Price (EUR)
61,60
Each
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
61,60
Each
Chemical Identifiers
CAS144104-59-6
IUPAC Name(1H-indol-5-yl)boronic acid
Molecular FormulaC8H8BNO2
InChI KeyVHADYSUJZAPXOW-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige
Appearance (Form)Powder and/or chunks
Titration with NaOH>=96.0 %
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only