3,4-Methylenedioxyphenylboronic acid, 98%
3,4-Methylenedioxyphenylboronic acid, 98%
3,4-Methylenedioxyphenylboronic acid, 98%
Thermo Scientific Chemicals

3,4-Methylenedioxyphenylboronic acid, 98%

CAS: 94839-07-3 | C7H7BO4 | 165.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 359580010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS94839-07-3
IUPAC Name(2H-1,3-benzodioxol-5-yl)boronic acid
Molecular FormulaC7H7BO4
InChI KeyCMHPUBKZZPSUIQ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH97.5 to 106.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only