4-Thiouracil, 97%
4-Thiouracil, 97%
4-Thiouracil, 97%
Thermo Scientific Chemicals

4-Thiouracil, 97%

CAS: 591-28-6 | C4H4N2OS | 128.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3599300101 gGlass bottle
Catalog number 359930010
Price (EUR)
103,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
103,00
Each
Chemical Identifiers
CAS591-28-6
IUPAC Name4-sulfanylidene-1,2,3,4-tetrahydropyrimidin-2-one
Molecular FormulaC4H4N2OS
InChI KeyOVONXEQGWXGFJD-UHFFFAOYSA-N
SMILESO=C1NC=CC(=S)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Appearance of solution(5% sol. in 1N NaOH) Clear and faint yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only