2-Methyl-2-pentenal, 97%
2-Methyl-2-pentenal, 97%
2-Methyl-2-pentenal, 97%
Thermo Scientific Chemicals

2-Methyl-2-pentenal, 97%

CAS: 623-36-9 | C6H10O | 98.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 359960250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS623-36-9
IUPAC Name(2E)-2-methylpent-2-enal
Molecular FormulaC6H10O
InChI KeyIDEYZABHVQLHAF-GQCTYLIASA-N
SMILESCCC=C(/C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.4470 to 1.4550 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only